2,3-Dibromo-5,6,7,8-tetrahydroimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

114786-1G
102894-700EA 958.46 USD
102894-700
2,3-Dibromo-5,6,7,8-tetrahydroimidazo[1,2-a]pyridine ≥97%
2,3-Dibromo-5,6,7,8-tetrahydroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₈Br₂N₂
MW: 279.97 g/mol
MDL Number: MFCD27922699
CAS Number: 156817-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR