Methyl-2-morpholino-2-phenylacetate hydrochloride ≥97%

Supplier: Matrix Scientific

114713-1G
102894-580EA 813.85 USD
102894-580
Methyl-2-morpholino-2-phenylacetate hydrochloride ≥97%
Methyl-2-morpholino-2-phenylacetate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MW: 271.75 g/mol
MDL Number: MFCD28099762
CAS Number: 27594-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR