Ethyl-1-cyclopentylpiperidine-4-carboxylate ≥97%

Supplier: Matrix Scientific

114708-1G
102894-570EA 641.54 USD
102894-570
Ethyl-1-cyclopentylpiperidine-4-carboxylate ≥97%
Ethyl-1-cyclopentylpiperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₃NO₂
MDL Number: MFCD16160131
CAS Number: 733783-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR