Methyl-3-(2-aminopyrimidin-4-yl)benzoate ≥97%

Supplier: Matrix Scientific

114641-1G
102894-412EA 813.85 USD
102894-412
Methyl-3-(2-aminopyrimidin-4-yl)benzoate ≥97%
Methyl-3-(2-aminopyrimidin-4-yl)benzoate

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₂
MDL Number: MFCD28099750
CAS Number: 579476-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR