Methyl-4-(2-aminopyrimidin-4-yl)benzoate ≥97%

Supplier: Matrix Scientific

114640-1G
102894-410EA 813.85 USD
102894-410
Methyl-4-(2-aminopyrimidin-4-yl)benzoate ≥97%
Methyl-4-(2-aminopyrimidin-4-yl)benzoate

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₂
MW: 229.24 g/mol
MDL Number: MFCD28099749
CAS Number: 1706430-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR