6-Acetamidohexanoyl chloride ≥97%

Supplier: Matrix Scientific

114613-1G
102898-422EA 813.85 USD
102898-422
6-Acetamidohexanoyl chloride ≥97%
6-Acetamidohexanoyl chloride

Hazard: Irritant

Formula: C₈H₁₄ClNO₂
MDL Number: MFCD24552402
CAS Number: 43218-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR