3-Hydroxypyrazine-2-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114557-1G
102894-286EA 958.46 USD
102894-286
3-Hydroxypyrazine-2-carbonyl chloride ≥97%
3-Hydroxypyrazine-2-carbonyl chloride

Hazard: Irritant

Formula: C₅H₃ClN₂O₂
MDL Number: MFCD18416934
CAS Number: 98136-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR