2-Butyl-5-chloro-1H-imidazole-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
114533-1G
102894-240EA
958.46
USD
102894-240
2-Butyl-5-chloro-1H-imidazole-4-carboxylic acid ≥97%
2-Butyl-5-chloro-1H-imidazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₈H₁₁ClN₂O₂ |
MDL Number:
MFCD06203325 CAS Number: 149968-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...