6-(2-Piperidin-4-ylethoxy)pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

114521-1G
102894-216EA 958.46 USD
102894-216
6-(2-Piperidin-4-ylethoxy)pyridine-2-carboxylic acid ≥97%
6-(2-Piperidin-4-ylethoxy)pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD28099737
CAS Number: 1706438-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR