2-(2-Piperidin-4-ylethoxy)isonicotinic acid ≥97%

Supplier: Matrix Scientific

114520-1G
102894-214EA 958.46 USD
102894-214
2-(2-Piperidin-4-ylethoxy)isonicotinic acid ≥97%
2-(2-Piperidin-4-ylethoxy)isonicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD28099736
CAS Number: 1706430-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR