6-(2-Piperidin-4-ylethoxy)nicotinic acid ≥97%

Supplier: Matrix Scientific

114519-1G
102894-212EA 958.46 USD
102894-212
6-(2-Piperidin-4-ylethoxy)nicotinic acid ≥97%
6-(2-Piperidin-4-ylethoxy)nicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD28099735
CAS Number: 1706462-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR