5-Methoxynicotinoyl chloride ≥95%

Supplier: Matrix Scientific

114504-1G
102894-180EA 1175.38 USD
102894-180
5-Methoxynicotinoyl chloride ≥95%
5-Methoxynicotinoyl chloride

Hazard: Irritant

Formula: C₇H₆ClNO₂
MDL Number: MFCD28099734
CAS Number: 1123782-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR