4-(cis-2,6-Dimethylmorpholino)benzoic acid ≥97%

Supplier: Matrix Scientific

114451-1G
102894-066EA 958.46 USD
102894-066
4-(cis-2,6-Dimethylmorpholino)benzoic acid ≥97%
4-(cis-2,6-Dimethylmorpholino)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MDL Number: MFCD28099724
CAS Number: 1544684-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR