6,8-Difluoro[1,2,4]triazolo[4,3-a]pyridin-3-amine hydrobromide ≥97%
Supplier: Matrix Scientific
114390-1G
102893-930EA
1707.69
USD
102893-930
6,8-Difluoro[1,2,4]triazolo[4,3-a]pyridin-3-amine hydrobromide ≥97%
6,8-Difluoro[1,2,4]triazolo[4,3-a]pyridin-3-amine hydrobromide
Hazard: Irritant
Formula:
C₆H₅BrF₂N₄ MW: 251.04 g/mol |
MDL Number:
MFCD28099723 CAS Number: 1706463-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...