N-(1,2,3,4-Tetrahydronaphthalen-1-yl)-1H-pyrazol-4-amine ≥97%

Supplier: Matrix Scientific

114376-1G
102893-902EA 1443.08 USD
102893-902
N-(1,2,3,4-Tetrahydronaphthalen-1-yl)-1H-pyrazol-4-amine ≥97%
N-(1,2,3,4-Tetrahydronaphthalen-1-yl)-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₃H₁₅N₃
MDL Number: MFCD12133682
CAS Number: 1154174-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR