(4-Bromo-3-fluorophenyl)acetyl chloride ≥97%

Supplier: Matrix Scientific

114343-1G
102893-834EA 1298.46 USD
102893-834
(4-Bromo-3-fluorophenyl)acetyl chloride ≥97%
(4-Bromo-3-fluorophenyl)acetyl chloride

Hazard: Irritant

Formula: C₈H₅BrClFO
MDL Number: MFCD28099717
CAS Number: 1360868-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR