(3-Fluoro-4-methoxyphenyl)acetyl chloride ≥95%

Supplier: Matrix Scientific

114325-1G
102893-796EA 958.46 USD
102893-796
(3-Fluoro-4-methoxyphenyl)acetyl chloride ≥95%
(3-Fluoro-4-methoxyphenyl)acetyl chloride

Hazard: Irritant

Formula: C₉H₈ClFO₂
MDL Number: MFCD12403405
CAS Number: 1207516-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR