2-Oxo-4-phenylbutyroyl chloride ≥97%

Supplier: Matrix Scientific

114312-1G
102898-406EA 813.85 USD
102898-406
2-Oxo-4-phenylbutyroyl chloride ≥97%
2-Oxo-4-phenylbutyroyl chloride

Hazard: Irritant

Formula: C₁₀H₉ClO₂
MDL Number: MFCD28099712
CAS Number: 1706437-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR