2-Amino-4-methylbenzoyl chloride ≥97%

Supplier: Matrix Scientific

114300-1G
102893-768EA 1069.23 USD
102893-768
2-Amino-4-methylbenzoyl chloride ≥97%
2-Amino-4-methylbenzoyl chloride

Hazard: Irritant

Formula: C₈H₈ClNO
MDL Number: MFCD18389823
CAS Number: 1261522-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR