3,4-Difluoro-2-methoxybenzoyl chloride ≥97%

Supplier: Matrix Scientific

114290-1G
102893-748EA 958.46 USD
102893-748
3,4-Difluoro-2-methoxybenzoyl chloride ≥97%
3,4-Difluoro-2-methoxybenzoyl chloride

Hazard: Irritant

Formula: C₈H₅ClF₂O₂
MDL Number: MFCD28099709
CAS Number: 1706461-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR