2-Bromo-3-(trifluoromethyl)benzenesulfonamide ≥97%

Supplier: Matrix Scientific

114192-1G
102893-528EA 1443.08 USD
102893-528
2-Bromo-3-(trifluoromethyl)benzenesulfonamide ≥97%
2-Bromo-3-(trifluoromethyl)benzenesulfonamide

Hazard: Irritant

Formula: C₇H₅BrF₃NO₂S
MDL Number: MFCD03425152
CAS Number: 886762-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR