2-(4-tert-Butylbenzylthio)ethylamine ≥97%

Supplier: Matrix Scientific

114144-10G 114144-1G
102893-434EA 156.92 USD
102893-434 102893-436
2-(4-tert-Butylbenzylthio)ethylamine ≥97%
2-(4-tert-Butylbenzylthio)ethylamine

Hazard: Irritant

Formula: C₁₃H₂₁NS
MDL Number: MFCD01764883
CAS Number: 260802-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR