4-Bromo-3-cyclopropyl-1-phenyl-1H-pyrazole-5-amine ≥97%

Supplier: Matrix Scientific

114074-1G
102893-256EA 813.85 USD
102893-256
4-Bromo-3-cyclopropyl-1-phenyl-1H-pyrazole-5-amine ≥97%
4-Bromo-3-cyclopropyl-1-phenyl-1H-pyrazole-5-amine

Hazard: Irritant

Formula: C₁₂H₁₂BrN₃
MDL Number: MFCD20233391
CAS Number: 1349709-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR