5-Chloro-2-(4-nitro-1H-pyrazol-1-yl)pyridine ≥97%

Supplier: Matrix Scientific

114063-1G
102893-234EA 813.85 USD
102893-234
5-Chloro-2-(4-nitro-1H-pyrazol-1-yl)pyridine ≥97%
5-Chloro-2-(4-nitro-1H-pyrazol-1-yl)pyridine

Hazard: Irritant

Formula: C₈H₅ClN₄O₂
MDL Number: MFCD22300562
CAS Number: 1424857-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR