1-Bromo-2-methyl-3,5-bis(trifluoromethyl)benzene ≥95%
Supplier: Matrix Scientific
114058-1G
102893-228EA
813.85
USD
102893-228
1-Bromo-2-methyl-3,5-bis(trifluoromethyl)benzene ≥95%
1-Bromo-2-methyl-3,5-bis(trifluoromethyl)benzene
Hazard: Irritant
Formula:
C₉H₅BrF₆ |
MDL Number:
MFCD01631703 CAS Number: 251651-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...