Diethyl cyclobutylidenemalonate ≥97%
Supplier: Matrix Scientific
114017-1G
102893-146EA
884.62
USD
102893-146
Diethyl cyclobutylidenemalonate ≥97%
Diethyl cyclobutylidenemalonate
Hazard: Irritant
Formula:
C₁₁H₁₆O₄ |
MDL Number:
MFCD22575251 CAS Number: 41589-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...