Ethyl-4-(cyclopropylamino)-2-(methylthio)pyrimidine-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(cyclopropylamino)-2-(methylthio)pyrimidine-5-carboxylate

114001-1G
102893-112EA 813.85 USD
102893-112
Ethyl-4-(cyclopropylamino)-2-(methylthio)pyrimidine-5-carboxylate ≥97%
Ethyl-4-(cyclopropylamino)-2-(methylthio)pyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂S
MW: 253.33 g/mol
MDL Number: MFCD27955205
CAS Number: 651734-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR