Methyl-2-chloro-5-fluoro-6-piperazin-1-ylnicotinate ≥97%

Supplier: Matrix Scientific

113961-1G
102898-094EA 813.85 USD
102898-094
Methyl-2-chloro-5-fluoro-6-piperazin-1-ylnicotinate ≥97%
Methyl-2-chloro-5-fluoro-6-piperazin-1-ylnicotinate

Hazard: Irritant

Formula: C₁₁H₁₃ClFN₃O₂
MDL Number: MFCD26099155
CAS Number: 1706436-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR