Methyl-3,5-difluoro-2-(2-methoxy-2-oxoethoxy)benzoate ≥97%

Supplier: Matrix Scientific

113948-1G
102898-074EA 738.46 USD
102898-074
Methyl-3,5-difluoro-2-(2-methoxy-2-oxoethoxy)benzoate ≥97%
Methyl-3,5-difluoro-2-(2-methoxy-2-oxoethoxy)benzoate

Hazard: Irritant

Formula: C₁₁H₁₀F₂O₅
MDL Number: MFCD26099150
CAS Number: 1706451-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR