5-Hydroxy-2-nitrobenzoyl chloride ≥95%

Supplier: Matrix Scientific

113874-1G
102898-122EA 813.85 USD
102898-122
5-Hydroxy-2-nitrobenzoyl chloride ≥95%
5-Hydroxy-2-nitrobenzoyl chloride

Hazard: Irritant

Formula: C₇H₄ClNO₄
MDL Number: MFCD18397351
CAS Number: 1261617-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR