4-(Trifluoromethyl)nicotinic acid-1-oxide ≥97%

Supplier: Matrix Scientific

113834-1G
102892-882EA 958.46 USD
102892-882
4-(Trifluoromethyl)nicotinic acid-1-oxide ≥97%
4-(Trifluoromethyl)nicotinic acid-1-oxide

Hazard: Irritant

Formula: C₇H₄F₃NO₃
MDL Number: MFCD06208787
CAS Number: 178748-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR