1H-Pyrrolo[2,3-b]pyridine-3-carbonyl chloride ≥96%

Supplier: Matrix Scientific

113791-1G
102892-794EA 958.46 USD
102892-794
1H-Pyrrolo[2,3-b]pyridine-3-carbonyl chloride ≥96%
1H-Pyrrolo[2,3-b]pyridine-3-carbonyl chloride

Hazard: Irritant

Formula: C₈H₅ClN₂O
MDL Number: MFCD18414669
CAS Number: 721882-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR