4-Bromo-3-methoxybenzoyl chloride ≥97%

Supplier: Matrix Scientific

113652-1G
102897-980EA 813.85 USD
102897-980
4-Bromo-3-methoxybenzoyl chloride ≥97%
4-Bromo-3-methoxybenzoyl chloride

Hazard: Irritant

Formula: C₈H₆BrClO₂
MDL Number: MFCD18391883
CAS Number: 56256-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR