3-(2-Bromo-4,5-difluorophenoxy)propanoic acid ≥97%

Supplier: Matrix Scientific

113626-1G
102897-930EA 690.77 USD
102897-930
3-(2-Bromo-4,5-difluorophenoxy)propanoic acid ≥97%
3-(2-Bromo-4,5-difluorophenoxy)propanoic acid

Hazard: Irritant

Formula: C₉H₇BrF₂O₃
MDL Number: MFCD26099136
CAS Number: 911826-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR