(5-[(Methylamino)methyl]furan-2-yl)methanol ≥97%

Supplier: Matrix Scientific

113594-1G
102892-508EA 958.46 USD
102892-508
(5-[(Methylamino)methyl]furan-2-yl)methanol ≥97%
(5-[(Methylamino)methyl]furan-2-yl)methanol

Hazard: Irritant

Formula: C₇H₁₁NO₂
MDL Number: MFCD11046474
CAS Number: 66357-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR