N-{[6-(1H-Imidazol-1-yl)pyridin-3-yl]methyl}-N-methylamine ≥97%
Supplier: Matrix Scientific
113554-1G
102897-884EA
1333.85
USD
102897-884
N-{[6-(1H-Imidazol-1-yl)pyridin-3-yl]methyl}-N-methylamine ≥97%
N-{[6-(1H-Imidazol-1-yl)pyridin-3-yl]methyl}-N-methylamine
Hazard: Irritant
Formula:
C₁₀H₁₂N₄ |
MDL Number:
MFCD16703994 CAS Number: 1247863-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...