2-Oxa-spiro[3.3]heptan-6-one

Supplier: Matrix Scientific

112981-1G
102891-316EA 2850.77 USD
102891-316
2-Oxa-spiro[3.3]heptan-6-one
2-Oxa-spiro[3.3]heptan-6-one

Hazard: Irritant

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD22415239
CAS Number: 1339892-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR