3-(3-Bromo-4-hydroxyphenyl)-2-(hydroxyimino)propanoic acid ≥97%

Supplier: Matrix Scientific

087222-500MG
102852-958EA 600 USD
102852-958
3-(3-Bromo-4-hydroxyphenyl)-2-(hydroxyimino)propanoic acid ≥97%
3-(3-Bromo-4-hydroxyphenyl)-2-(hydroxyimino)propanoic acid

Hazard: Irritant

Formula: C₉H₈BrNO₄
MDL Number: MFCD22573521
CAS Number: 146884-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR