3-Methyl-5-(2-(methylthio)phenyl)-5-oxovaleric acid ≥97%

Supplier: Matrix Scientific

087053-1G 087053-2G 087053-5G
102852-214EA 490.77 USD
102852-214 102852-216 102852-218
3-Methyl-5-(2-(methylthio)phenyl)-5-oxovaleric acid ≥97%
3-Methyl-5-(2-(methylthio)phenyl)-5-oxovaleric acid

Hazard: Irritant

Formula: C₁₃H₁₆O₃S
MDL Number: MFCD09801818
CAS Number: 951892-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR