5-(3-Fluorobenzoyl)-2-methylpyridine ≥97%

Supplier: Matrix Scientific

086655-1G 086655-5G 086655-2G
102849-838EA 795.38 USD
102849-838 102849-842 102849-840
5-(3-Fluorobenzoyl)-2-methylpyridine ≥97%
5-(3-Fluorobenzoyl)-2-methylpyridine

Hazard: Irritant

Formula: C₁₃H₁₀FNO
MDL Number: MFCD13153214
CAS Number: 1187168-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR