3-(3,4,5-Trifluorobenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086652-5G 086652-2G 086652-1G
102849-824EA 2172.31 USD
102849-824 102849-822 102849-820
3-(3,4,5-Trifluorobenzoyl)pyridine ≥97%
3-(3,4,5-Trifluorobenzoyl)pyridine

Hazard: Irritant

Formula: C₁₂H₆F₃NO
MDL Number: MFCD13152937
CAS Number: 1187171-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR