2-Benzoyl-6-methoxypyridine ≥97%

Supplier: Matrix Scientific

086645-1G 086645-2G 086645-5G
102849-782EA 2172.31 USD
102849-782 102849-780 102849-778
2-Benzoyl-6-methoxypyridine ≥97%
2-Benzoyl-6-methoxypyridine

Hazard: Irritant

Formula: C₁₃H₁₁NO₂
MDL Number: MFCD13152835
CAS Number: 29082-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR