trans-2-(2-Thiomethylbenzoyl)cyclopentane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086443-2G
102848-720EA 3076.92 USD
102848-720
trans-2-(2-Thiomethylbenzoyl)cyclopentane-1-carboxylic acid ≥95%
trans-2-(2-Thiomethylbenzoyl)cyclopentane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₆O₃S
Melting Pt: 135…137 °C
MDL Number: MFCD01311238
CAS Number: 1134700-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR