cis-2-(2-Thiomethylbenzoyl)cyclopentane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086438-1G 086438-5G
102848-688EA 1203.08 USD
102848-688 102848-692
cis-2-(2-Thiomethylbenzoyl)cyclopentane-1-carboxylic acid ≥95%
cis-2-(2-Thiomethylbenzoyl)cyclopentane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₆O₃S
MDL Number: MFCD01311197
CAS Number: 1134527-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR