Ethyl-2-(thiomethyl)benzoylformate ≥97%

Supplier: Matrix Scientific

086415-2G
102848-552EA 827.69 USD
102848-552
Ethyl-2-(thiomethyl)benzoylformate ≥97%
Ethyl-2-(thiomethyl)benzoylformate

Hazard: Irritant

Formula: C₁₁H₁₂O₃S
MDL Number: MFCD09801441
CAS Number: 951888-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR