Ethyl-3-fluoro-2-methylbenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(3-fluoro-2-methylphenyl)-2-oxoacetate

086400-1G 086400-5G
102848-460EA 530.77 USD
102848-460 102848-464
Ethyl-3-fluoro-2-methylbenzoylformate ≥97%
Ethyl-3-fluoro-2-methylbenzoylformate

Hazard: Irritant/Refrigerate

Formula: C₁₁H₁₁FO₃
MW: 210.21 g/mol
MDL Number: MFCD09801424
CAS Number: 951888-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR