Ethyl-4-(1,3-dioxan-2-yl)-3-fluorobenzoylformate ≥97%

Supplier: Matrix Scientific

086390-2G
102848-402EA 827.69 USD
102848-402
Ethyl-4-(1,3-dioxan-2-yl)-3-fluorobenzoylformate ≥97%
Ethyl-4-(1,3-dioxan-2-yl)-3-fluorobenzoylformate

Hazard: Irritant

Formula: C₁₄H₁₅FO₅
MDL Number: MFCD09801415
CAS Number: 951888-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR