Ethyl-3-chloro-4-methylbenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(3-chloro-4-methylphenyl)-2-oxoacetate

086369-5G 086369-1G
102848-274EA 530.77 USD
102848-274 102848-278
Ethyl-3-chloro-4-methylbenzoylformate ≥97%
Ethyl-3-chloro-4-methylbenzoylformate

Hazard: Irritant

Formula: C₁₁H₁₁ClO₃
MW: 226.66 g/mol
MDL Number: MFCD09801397
CAS Number: 951888-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR