3-(4-Bromo-2-fluorophenyl)pyridine ≥97%

Supplier: Matrix Scientific

086307-1G 086307-5G
102847-914EA 592.31 USD
102847-914 102847-918
3-(4-Bromo-2-fluorophenyl)pyridine ≥97%
3-(4-Bromo-2-fluorophenyl)pyridine

Hazard: Irritant

Formula: C₁₁H₇BrFN
Melting Pt: 51…52 °C
MDL Number: MFCD13152460
CAS Number: 1187163-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR