3-(4-Ethylphenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086245-1G 086245-5G
102847-548EA 387.69 USD
102847-548 102847-552
3-(4-Ethylphenyl)-1-propene ≥97%
3-(4-Ethylphenyl)-1-propene

Hazard: Irritant

Formula: C₁₁H₁₄
MDL Number: MFCD06201189
CAS Number: 18640-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR